2-Methoxy-5-(trifluoromethyl)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methoxy-5-(trifluoromethyl)benzeneboronic acid
A230790-1G
A230790-25G
A230790-5G
76692-058EA
16
USD
76692-058
76692-060
76692-062
2-Methoxy-5-(trifluoromethyl)phenylboronic acid 98%
2-Methoxy-5-(trifluoromethyl)phenylboronic acid
Caution: for Research Use Only
Formula:
C₈H₈BF₃O₃ |
MDL Number:
MFCD04039222 CAS Number: 240139-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...