2-Methoxy-5-(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-5-(trifluoromethyl)benzeneboronic acid

A230790-1G A230790-25G A230790-5G
76692-058EA 16 USD
76692-058 76692-060 76692-062
2-Methoxy-5-(trifluoromethyl)phenylboronic acid 98%
2-Methoxy-5-(trifluoromethyl)phenylboronic acid

Caution: for Research Use Only

Formula: C₈H₈BF₃O₃
MDL Number: MFCD04039222
CAS Number: 240139-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR