Benzoxazole 98%
Supplier: Ambeed
|
Warning
|
A224260-100G
A224260-10G
A224260-25G
A224260-500G
A224260-5G
76691-892EA
40
USD
76691-892
76691-894
76691-896
76691-898
76691-900
Benzoxazole 98%
Benzoxazole
Caution: for Research Use Only
Formula:
C₇H₅NO |
MDL Number:
MFCD00005765 CAS Number: 273-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...