Sodium dehydracetate 98%

Supplier: Ambeed
Warning

Synonyms: Dehydracetic acid sodium salt, Dehydroacetic acid sodium salt, Sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide, Sodium dehydroacetate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate

A214483-1000G A214483-100G A214483-500G
76691-588EA 32 USD
76691-588 76691-590 76691-592
Sodium dehydracetate 98%
Sodium dehydracetate

Caution: for Research Use Only

Formula: C₈H₇NaO₄
MDL Number: MFCD00040583
CAS Number: 4418-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR