Sodium dehydracetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dehydracetic acid sodium salt, Dehydroacetic acid sodium salt, Sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide, Sodium dehydroacetate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate
A214483-1000G
A214483-100G
A214483-500G
76691-588EA
32
USD
76691-588
76691-590
76691-592
Sodium dehydracetate 98%
Sodium dehydracetate
Caution: for Research Use Only
Formula:
C₈H₇NaO₄ |
MDL Number:
MFCD00040583 CAS Number: 4418-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...