Bis(pentamethylcyclopentadienyl)iron(II) 98%

Supplier: Ambeed
Warning

Synonyms: Bis(pentamethylcyclopentadienyl)iron, Decamethylferrocene

A213221-10G A213221-1G A213221-250MG A213221-5G
76691-552EA 332 USD
76691-552 76691-554 76691-556 76691-558
Bis(pentamethylcyclopentadienyl)iron(II) 98%
Bis(pentamethylcyclopentadienyl)iron (II)

Caution: for Research Use Only

Formula: C₂₀H₃₀Fe
MDL Number: MFCD00058736
CAS Number: 12126-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR