D-Biotin-N-succinimidyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide

A209315-100MG A209315-1MG A209315-250MG A209315-1G A209315-5G
103817-934EA 21 USD
103817-934 103848-620 76691-416 76691-418 76691-420 77086-960 77086-962
D-Biotin-N-succinimidyl ester 97%
D-Biotin-N-succinimidyl ester

Caution: for Research Use Only

Formula: C₁₄H₁₉N₃O₅S
Melting Pt: 212…214 °C
Storage Temperature: Freezer
MDL Number: MFCD00078531
CAS Number: 35013-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR