Benzyl alcohol 98%

Supplier: Ambeed
Warning

A194236-1000G A194236-100G A194236-500G
76690-988EA 13 USD
76690-988 76690-990 76690-992
Benzyl alcohol 98%
Benzyl alcohol

Caution: for Research Use Only

Formula: C₆H₅CH₂OH
MDL Number: MFCD00004599
CAS Number: 100-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR