Tetraethylene glycol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol
A148598-1000G
A148598-100G
A148598-25G
A148598-500G
76689-262EA
27
USD
76689-262
76689-264
76689-266
76689-268
Tetraethylene glycol 98%
Tetraethylene glycol
Caution: for Research Use Only
Formula:
C₈H₁₈O₅ MW: 194.23 g/mol |
MDL Number:
MFCD00002879 CAS Number: 112-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...