Boc-L-Nle(6-OH) 95%

Supplier: Ambeed
Warning

Synonyms: Boc-6-OH-Nle-OH, (S)-2-((tert-Butoxycarbonyl)amino)-6-hydroxyhexanoic acid

A139086-1G A139086-250MG A139086-5G
76688-950EA 57 USD
76688-950 76688-952 76688-954
Boc-L-Nle(6-OH) 95%
Boc-L-Nle(6-OH)

Caution: for Research Use Only

Formula: C₁₁H₂₁NO₅
MW: 247.3 g/mol
MDL Number: MFCD01862292
CAS Number: 77611-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR