Copper(II) gluconate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate
A135321-1000G
A135321-100G
A135321-25G
A135321-500G
76688-778EA
83
USD
76688-778
76688-780
76688-782
76688-784
Copper(II) gluconate 98%
Copper(II) gluconate
Caution: for Research Use Only
Formula:
C₁₂H₂₂CuO₁₄ |
MDL Number:
MFCD00075297 CAS Number: 527-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...