Chloro(1,5-cyclooctadiene)iridium(I) dimer 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
[Ir(cod)Cl]2, Chloro-1,5-cyclooctadiene iridium(I) dimer
A132084-1G
A132084-250MG
A132084-5G
A132084-100MG
76688-672EA
199
USD
76688-672
76688-674
76688-676
77566-562
Chloro(1,5-cyclooctadiene)iridium(I) dimer 97%
Chloro(1,5-cyclooctadiene)iridium (I) dimer
Caution: For research use only.
Formula:
C₁₆H₂₄Cl₂Ir₂ MW: 671.70 g/mol |
MDL Number:
MFCD00012414 CAS Number: 12112-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...