(R)-(-)-Hydratropic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid
A123519-100G
A123519-10G
A123519-25G
A123519-1G
A123519-5G
103800-412EA
6
USD
103800-412
103831-250
76688-220
76688-222
76688-224
77297-948
77312-530
(R)-(-)-Hydratropic acid 98%
(R)-(-)-Hydratropic acid
Caution: for Research Use Only
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 257 Melting Pt: 29…30 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063140 CAS Number: 7782-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...