(R)-(-)-Hydratropic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid

A123519-100G A123519-10G A123519-25G A123519-1G A123519-5G
103800-412EA 6 USD
103800-412 103831-250 76688-220 76688-222 76688-224 77297-948 77312-530
(R)-(-)-Hydratropic acid 98%
(R)-(-)-Hydratropic acid

Caution: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 257
Melting Pt: 29…30 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00063140
CAS Number: 7782-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR