Bis(diphenylphosphino)methane 98%

Supplier: Ambeed
Warning

Synonyms: [(1,1-Diphenylphosphino)methyl](diphenyl)phosphine, DPPM

A118894-100G A118894-10G A118894-25G A118894-500G A118894-1G A118894-5G
103799-126EA 5 USD
103799-126 103830-498 76688-022 76688-024 76688-026 76688-028 77297-634 77311-800
Bis(diphenylphosphino)methane 98%
Bis(diphenylphosphino)methane

Caution: for Research Use Only

Formula: C₂₅H₂₂P₂
Boiling Pt: 497.2 °C (760 mm Hg)
Melting Pt: 118…119 °C
Flash Pt: 270.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00003537
CAS Number: 2071-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR