Bicine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

A114701-1000G A114701-100G A114701-10G A114701-25G A114701-500G
76687-742EA 74 USD
76687-742 76687-744 76687-746 76687-748 76687-750
Bicine 95%
Bicine

Caution: for Research Use Only

Formula: C₆H₁₃NO₄
MDL Number: MFCD00004295
CAS Number: 150-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR