Globe Glass™ Griffin Style Beakers, Low Form, Heavy Duty
Supplier: Globe Scientific
8020250 8020400 8020600 8021000 8022000 8024000
76683-710BX 138.49 USD
76683-710 76683-684 76683-716 76683-720 76683-692 76683-718
Globe Glass™ Griffin Style Beakers, Low Form, Heavy Duty
Beakers
Uniform wall thickness, flat bottoms and low expansion 3.3 Borosilicate glass make Globe Glass™ heavy duty, low form Griffin style beakers strong and durable.
  • Made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass
  • GlassGuard™ packaging system protects against transit damage
  • Dual metric scale graduations
  • Durable white enamel printing
  • Large white marking area
  • Formed pour spout for easy pouring
Thicker walls improve mechanical strength to protect against breakage even with the most rigorous use. Beaded rims and formed spouts facilitate accurate and steady pouring of liquids. Globe Glass low form Griffin style beakers are available in sizes from 250 to 4000 ml.
Certifications: Meet ASTM E960 Type II Specifications.