Normal Human Serum
Supplier: GeminiBio
100-110-100
76683-640EA 158.87 USD
76683-640
Normal Human Serum
Sera (Serum)
Normal human serum is collected from healthy male and female donors of all ABO serotypes at FDA-licensed facilities in the United States. This material is defibrinated from source plasma. All donor units are tested for viral markers and found to be non-reactive.
  • 0.1 µm sterile-filtered
Delivery information: Ships on dry ice