GemCell Plus™ U.S. Human Serum AB - Xeno-Free
Supplier: GeminiBio
100-912-100 100-912G-100
76683-634EA 518.13 USD
76683-634 76683-622
GemCell Plus™ U.S. Human Serum AB - Xeno-Free
Sera (Serum)
GemCell Plus™ Xeno-Free Human Serum is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material goes through additional rigorous testing and contains therapeutic grade human thrombin. All donor units are tested for viral markers and found to be non-reactive.
  • Developed without the use of animal material
  • Gamma irradiation available