VWR® Orbital Shaker With Magnetic Drive For Use in CO₂ Incubators
Supplier: VWR International
OSC-26M-VWR
76683-266EA 4313.75 USD
76683-266
VWR® Orbital Shaker With Magnetic Drive For Use in CO₂ Incubators
Shakers and Mixers Orbital Shakers
This versatile, reliable orbital shaker has been developed to help economically scale-up cell production in your existing CO₂ incubator.
  • Sealed electronics to minimize heat disruption to ensure conditions in CO₂ chamber not effected
  • LED display on remote control shows both set and actual parameters without opening the door
  • Alarm function alerts user when timer reaches zero
  • Orbital shaker with magnetic drive system for low vibration, and long service life, no belts
  • Ideal for use in CO₂, standard and refrigerated incubators up to 20% CO₂ and 90% humidity
  • Wide range of accessories to accommodate various sample holders
Shaker is easy to remove for clean/disinfecting the CO₂ chamber to protect cells. Additional platforms to suit the application are available as optional accessories.
Certifications: CE, UL, cUL Certified
Accessories information: Capacity: 30×25/50 ml, 15×100/150/200/250 ml, 9×500 ml, 6×1 L, 4×2 L, 2×3 L or 1×5 L flasks.
Delivery information: Supplied with remote control, platform with rubber mat (76683-262), universal tray (installed), power supply/cable and instruction manual. Unit has a 2 year warranty.