TempoDual™ ATPMito Rapid Dual Biosensor Cytotoxicity Assay
Supplier: TEMPO BIOSCIENCE, INC
403
76682-956EA 929.58 USD
76682-956
TempoDual™ ATPMito Rapid Dual Biosensor Cytotoxicity Assay
Assays Cellular Assays
Dual TempoATP-TempoMito Biosensor Cytotoxicity Assay.
  • Cryovial. ~1 million cells/ml
Product Type:
human cells
Long-Term Storage:
liquid nitrogen
Hazard Identifications:
Non hazardous
Non infectious
Non dangerous
Not for use in humans or animals
For Research & Development (R&D) only
Certifications: BSL2