L-Lysine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH

A1371828-1000G A1371828-500G
76682-426EA 44.56 USD
76682-426 76682-428 76682-430
L-Lysine 98%
L-Lysine

Caution: for Research Use Only

Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
MDL Number: MFCD00064433
CAS Number: 56-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR