(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic Acid, H-D-Tic-OH, D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

A130899-100G A130899-10G A130899-1G A130899-250MG A130899-25G A130899-5G
76681-626EA 140 USD
76681-626 76681-628 76681-630 76681-632 76681-634 76681-636
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Caution: for Research Use Only

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD00144038
CAS Number: 103733-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR