Dichloro(1,5-cyclooctadiene)palladium(II) 98+%

Supplier: Ambeed
Warning

Synonyms: 1,5-Cyclooctadienepalladium(II) Dichloride

A1233131-100MG
76679-914EA 19 USD
76679-914 76679-916 76679-918
Dichloro(1,5-cyclooctadiene)palladium(II) 98+%
Dichloro(1,5-cyclooctadiene)palladium(II)

Caution: for Research Use Only

Formula: C₈H₁₂Cl₂Pd
MDL Number: MFCD00012412
CAS Number: 12107-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR