Palladium(II) acetate 99.9%

Supplier: Ambeed
Danger

Synonyms: Palladium diacetate, Palladium (II) acetate

A1194522-10G A1194522-1G A1194522-250MG A1194522-25G A1194522-5G
76677-244EA 725 USD
76677-244 76677-246 76677-248 76677-250 76677-252
Palladium(II) acetate 99.9%
Palladium (II) acetate

Caution: for Research Use Only

Formula: C₄H₆O₄Pd
MDL Number: MFCD00012453
CAS Number: 3375-31-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR