Bisindoylmaleimide X hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Bisindolylmaleimide X HCl, Ro 31-8425, 3-(8-(Aminomethyl)-6,7,8,9-tetrahydropyrido[1,2-a]indol-10-yl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione hydrochloride
A1159144-10MG
A1159144-25MG
A1159144-5MG
76675-470EA
223
USD
76675-470
76675-472
76675-474
Bisindoylmaleimide X hydrochloride 98%
Bisindoylmaleimide X hydrochloride
Caution: for Research Use Only
Formula:
C₂₆H₂₄N₄O₂·HCl |
MDL Number:
MFCD04972010 CAS Number: 145317-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...