Precellys® Evolution Touch Homogenizer
Supplier: BERTIN CORP
P002511-PEVT0-A.0
76670-674EA 15519.58 USD
76670-674
Precellys® Evolution Touch Homogenizer
Homogenizers Benchtop Homogenizers
The Precellys Evolution Touch is both flexible and efficient, giving users the ability to grind, homogenize or lyse any kind of sample.
  • Use five different tube volumes (from 0.3 to 15 ml) and prepare between 1 and 96 tubes at the same time
  • From 4500 to 10000 rpm (increments of 100 rpm)
  • Automatic tube locking system
  • The software allows users to save an unlimited number of protocols
  • Run up to 10 cycles
Easy to use with its new intuitive touchscreen interface, designed to be used with gloves, researchers can easily set up the Precellys Evolution Touch. The Precellys Evolution Touch puts the power of homogenization into the hands of researchers and scientists.
Certifications: E and CFR 47 part 15 certified.