Drosophila Olfactory Operant Conditioning Apparatus
Supplier: MAZE ENGINEERS
8501
76670-412EA 6533.34 USD
76670-412
Drosophila Olfactory Operant Conditioning Apparatus
Animal Supplies Animal Environmental Enrichment
The Drosophila Olfactory Operant Conditioning is an odor-aversion learning apparatus for Drosophila.
  • 2 odor chambers
  • T-Maze choice area
  • Copper testing tube
  • Vacuum lines
  • Secure clamp
The Drosophila Olfactory Operant Conditioning apparatus was first developed by Tully and Quinn (Tully and Quinn, 1985) using a combination of a T-Maze electric-shock reinforcement in a copper wire grid. The unit includes a clear, robust apparatus that includes a holding area and T-Maze choice areas leading into odor compartments.