Parallel Bars
Supplier: MAZE ENGINEERS
5007
76670-194EA 1320 USD
76670-194
Parallel Bars
Animal Supplies Animal Environmental Enrichment
The Parallel Bars task is used in measuring and determining motor capabilities.
  • Acrylic frame and metal rods
  • Easy to use, clean, assemble, and disassemble
The Parallel Bars apparatus uses two parallel rods suspended using wooden columns on either end of the rods. The distance between the two columns or the space between the two parallel rods can be altered to create different levels of difficulty.