Automated Zebrafish Y-Maze
Supplier: MAZE ENGINEERS
4874
76670-134EA 5320 USD
76670-134
Automated Zebrafish Y-Maze
Animal Supplies Animal Environmental Enrichment
The automated avoidance Zebrafish Y-Maze is an operant conditioning assay used to study avoidance behaviors in Zebrafish when presented with an aversive stimulus.
  • LCD screen placed below maze displays visual cues throughout all maze regions
  • Shock and sound stimuli also available
  • Custom coloring and customization available
The Automated Avoidance Zebrafish Y-Maze consists of a Y-Maze with an LCD screen placed below the maze that displays visual cues throughout all regions of the maze.
Ordering information: The Automated Zebrafish Y-Maze requires software to function (see accessory options). Software package is designed for use with the Automated Zebrafish Y-Maze as a standalone software package.