Di(trimethylolpropane) 98% (by GC), (Chunks or flakes)

Supplier: Ambeed
Warning

A1493480-100G A1493480-25G A1493480-500G A1493480-1000G
76665-954EA 13 USD
76665-954 76665-956 76665-958 77549-856
Di(trimethylolpropane) 98% (by GC), (Chunks or flakes)
Di(trimethylolpropane)
Formula: C₁₂H₂₆O₅
MDL Number: MFCD00192117
CAS Number: 23235-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR