4-Amino-2-fluorobenzoic acid 98%

Supplier: Ambeed
Danger

A1485706-100G A1485706-1G A1485706-25G A1485706-5G
76664-852EA 160 USD
76664-852 76664-854 76664-856 76664-858
4-Amino-2-fluorobenzoic acid 98%
4-Amino-2-fluorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13
MDL Number: MFCD01569397
CAS Number: 446-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR