2-Iodo-9,9-dimethylfluorene 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 2-Iodo-9,9-dimethyl-9H-fluorene

A1482763-25G A1482763-5G
76664-404EA 25 USD
76664-404 76664-406
2-Iodo-9,9-dimethylfluorene 98% (by HPLC)
2-Iodo-9,9-dimethylfluorene

Caution: for Research Use Only

Formula: C₁₅H₁₃I
MW: 320.17 g/mol
MDL Number: MFCD12024272
CAS Number: 144981-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR