(2-Chlorophenyl)acetic acid 99%

Supplier: Ambeed
Warning

Synonyms: 2-Chlorophenylacetic acid

A1482295-1000G A1482295-100G A1482295-25G A1482295-500G
76664-254EA 70 USD
76664-254 76664-256 76664-258 76664-260
(2-Chlorophenyl)acetic acid 99%
(2-Chlorophenyl)acetic acid

Caution: for Research Use Only

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
MDL Number: MFCD00004317
CAS Number: 2444-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR