Antazoline hydrochloride 99+%

Supplier: Ambeed
Warning

A1475906-100G A1475906-1G A1475906-25G A1475906-5G
76663-542EA 996 USD
76663-542 76663-544 76663-546 76663-548
Antazoline hydrochloride 99+%
Antazoline hydrochloride

Caution: for Research Use Only

Formula: C₁₇H₁₉N₃
MDL Number: MFCD00058145
CAS Number: 2508-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR