EDTA ferric monosodium salt 12.5-13.5% Fe

Supplier: Ambeed
Warning

A1473241-25G
76663-028EA 7 USD
76663-028
EDTA ferric monosodium salt 12.5-13.5% Fe
EDTA ferric monosodium salt

Caution: for Research Use Only

Formula: C₁₀H₁₂FeN₂NaO₈
MDL Number: MFCD00078215
CAS Number: 15708-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR