Sulfanilamide 99%

Supplier: Ambeed
Warning

Synonyms: 4-Aminobenzenesulfonamide, 4-Aminobenzenesulphonamide

A1458408-1000G A1458408-100G A1458408-500G
76661-860EA 32 USD
76661-860 76661-862 76661-864
Sulfanilamide 99%
Sulphanilamide

Caution: for Research Use Only

Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: 401
MDL Number: MFCD00007939
CAS Number: 63-74-1
Merck Index: 13,09010

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR