Chromeazurol S (Mordant blue 29), Analytical reagent (AR)

Supplier: Ambeed
Warning

A1455421-25G
76661-724EA 65 USD
76661-724
Chromeazurol S (Mordant blue 29), Analytical reagent (AR)
Chromeazurol S (Mordant blue 29)

Caution: for Research Use Only

Formula: C₂₃H₁₆Cl₂O₉S
MDL Number: MFCD00001615
CAS Number: 1667-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR