Fumaric acid 99%

Supplier: Ambeed
Warning

A1453559-100G A1453559-500G A1453559-1000G
76661-586EA 5 USD
76661-586 76661-588 77874-923
Fumaric acid 99%
Fumaric acid

Caution: for Research Use Only

Formula: C₄H₄O₄
MDL Number: MFCD00002700
CAS Number: 110-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR