Strontium chloride, anhydrous 99.5% (metals basis)

Supplier: Ambeed
Warning

A1453364-100G A1453364-25G A1453364-500G A1453364-5G
76661-570EA 44 USD
76661-570 76661-572 76661-574 76661-576
Strontium chloride, anhydrous 99.5% (metals basis)
Strontium chloride

Caution: for Research Use Only

Formula: Cl₂Sr
MW: 158.53
Boiling Pt: 1250
Melting Pt: 868
Density: 1.96
MDL Number: MFCD00011249
CAS Number: 10476-85-4
Merck Index: 12,09000

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR