Strontium chloride, anhydrous 99.5% (metals basis)
Supplier: Ambeed
|
Warning
|
A1453364-100G
A1453364-25G
A1453364-500G
A1453364-5G
76661-570EA
44
USD
76661-570
76661-572
76661-574
76661-576
Strontium chloride, anhydrous 99.5% (metals basis)
Strontium chloride
Caution: for Research Use Only
Formula:
Cl₂Sr MW: 158.53 Boiling Pt: 1250 Melting Pt: 868 Density: 1.96 |
MDL Number:
MFCD00011249 CAS Number: 10476-85-4 Merck Index: 12,09000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...