α-Bromo-4-iodotoulene 96%
Supplier: Ambeed
|
Danger
|
A1340201-10G
A1340201-1G
A1340201-25G
A1340201-5G
76658-026EA
23
USD
76658-026
76658-028
76658-030
76658-032
α-Bromo-4-iodotoulene 96%
α-Bromo-4-iodotoulene
Caution: for Research Use Only
Formula:
C₇H₆BrI |
MDL Number:
MFCD00209656 CAS Number: 16004-15-2 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...