α-Bromo-4-iodotoulene 96%

Supplier: Ambeed
Danger

A1340201-10G A1340201-1G A1340201-25G A1340201-5G
76658-026EA 23 USD
76658-026 76658-028 76658-030 76658-032
α-Bromo-4-iodotoulene 96%
α-Bromo-4-iodotoulene

Caution: for Research Use Only

Formula: C₇H₆BrI
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR