Lithium stearate 98%

Supplier: Ambeed
Warning

A920970-1000G A920970-100G A920970-25G A920970-500G
76655-390EA 39 USD
76655-390 76655-392 76655-394 76655-396
Lithium stearate 98%
Lithium stearate

Caution: for Research Use Only

Formula: C₁₈H₃₅LiO₂
MDL Number: MFCD00042032
CAS Number: 4485-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR