Polyethylene glycol monomethyl ether, M.W. ∼4000

Supplier: Ambeed
Warning

Synonyms: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH

A833141-100G A833141-1000G
76654-014EA 62 USD
76654-014 77784-175
Polyethylene glycol monomethyl ether, M.W. ∼4000
Polyethylene glycol monomethyl ether

Caution: for Research Use Only

Formula: CH₃(OCH₂CH₂)nOH
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR