Polyethylene glycol monomethyl ether, M.W. ∼2000
Supplier: Ambeed
|
Warning
|
Synonyms:
Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH
A641450-1000G
A641450-100G
A641450-10G
A641450-25G
A641450-500G
76650-518EA
62
USD
76650-518
76650-520
76650-522
76650-524
76650-526
Polyethylene glycol monomethyl ether, M.W. ∼2000
Polyethylene glycol monomethyl ether
Caution: for Research Use Only
Formula:
CH₃(OCH₂CH₂)nOH |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...