3'-Fluoroacetophenone 97%
Supplier: Ambeed
|
Warning
|
A497966-100G
A497966-10G
A497966-500G
A497966-5G
A497966-25G
76649-396EA
67
USD
76649-396
76649-398
76649-400
76649-402
76843-320
3'-Fluoroacetophenone 97%
3'-Fluoroacetophenone
Caution: for Research Use Only
Formula:
C₈H₇FO |
MDL Number:
MFCD00000338 CAS Number: 455-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...