3'-Fluoroacetophenone 97%

Supplier: Ambeed
Warning

A497966-100G A497966-10G A497966-500G A497966-5G A497966-25G
76649-396EA 67 USD
76649-396 76649-398 76649-400 76649-402 76843-320
3'-Fluoroacetophenone 97%

Caution: for Research Use Only

Formula: C₈H₇FO
MDL Number: MFCD00000338
CAS Number: 455-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR