Polyethylene glycol 400 Mn 380-420 ∼ Mn 400
Supplier: Ambeed
|
Warning
|
Synonyms:
PEG 400
A197578-1000G
A197578-100G
A197578-25G
A197578-500G
A197578-5G
76647-432EA
20
USD
76647-432
76647-434
76647-436
76647-438
76647-440
Polyethylene glycol 400 Mn 380-420 ∼ Mn 400
Polyethylene glycol 400
Caution: for Research Use Only
Formula:
H(OCH₂CH₂)nOH |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...