Polyethylene glycol 6,000 ∼ Mn 6000

Supplier: Ambeed
Warning

A172254-1000G A172254-100G A172254-25G A172254-500G
76647-224EA 20 USD
76647-224 76647-226 76647-228 76647-230
Polyethylene glycol 6,000 ∼ Mn 6000
Polyethylene glycol 6,000

Caution: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MDL Number: MFCD01779614
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR